Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294162.1 | NP_001281091.1 | 6 | 1724 | ||
NM_013569.2 | NP_038597.2 | 10 | 2752 | |||
GenBank mRNA | AF012868.1 | - | 10 | 2758 | ||
AF012869.1 | - | 6 | 1672 | |||
AF034762.1 | - | 6 | 1716 | |||
AK145127.1 | - | 1728 | ||||
AY374424.1 | - | 10 | 2437 | |||
BC051016.1 | - | 10 | 2583 | |||
BU707188.1 | - | 4 | 538 |