Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267593.1 | NP_001254522.1 | 2 | 1560 | ||
NM_010603.6 | NP_034733.4 | 3 | 1732 | |||
GenBank mRNA | AB035889.1 | - | 1 | 1192 | ||
AK017299.1 | - | 3 | 1733 | |||
AK032271.1 | - | 2 | 1562 | |||
AK162389.1 | - | 1 | 632 | |||
BC115970.1 | - | 2 | 1312 | 40106507 | ||
BC116677.1 | - | 2 | 1312 | 40106509 | ||
X80417.1 | - | 1 | 1297 |