Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252207.1 | NP_001239136.1 | 3 | 719 | ||
NM_001252208.1 | NP_001239137.1 | 2 | 696 | |||
NM_001252209.2 | NP_001239138.1 | 2 | 686 | |||
NM_001252210.2 | NP_001239139.1 | 2 | 667 | |||
NM_010604.3 | NP_034734.3 | 2 | 638 | |||
GenBank mRNA | AB016197.1 | - | 1 | 470 | ||
AK032192.1 | - | 2 | 652 | |||
AK045447.1 | - | 2 | 642 | |||
AK078905.1 | - | 2 | 639 | |||
AK128942.1 | - | 2 | 638 | |||
AK140249.1 | - | 2 | 649 | |||
AY377989.1 | - | 1 | 472 | |||
BC033442.1 | - | 3 | 694 | 4223640 | ||
BC092296.1 | - | 1 | 529 | 5064724 |