Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046778.1 | - | 2 | 843 | ||
BC068167.1 | - | 3 | 1722 | 30543982 | ||
BC103671.1 | - | 2 | 798 | |||
BC103673.1 | - | 2 | 798 | 40046971 | ||
BC103674.1 | - | 2 | 798 | 40046969 | ||
D45022.1 | - | 2 | 806 |