Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008429.2 | NP_032455.2 | 4 | 1193 | ||
GenBank mRNA | AF130860.1 | - | 4 | 1156 | ||
AK033970.1 | - | 4 | 1193 | |||
AK046084.1 | - | 4 | 1232 | |||
BC065161.1 | - | 3 | 994 | 5718431 | ||
U11860.1 | - | 4 | 1242 |