Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB000498.1 | - | 7 | 1063 | ||
AB000503.1 | - | 7 | 1063 | |||
AB000504.1 | - | 6 | 1621 | |||
AF490773.1 | - | 7 | 977 | |||
AK135059.1 | - | 6 | 635 | |||
AK139411.1 | - | 7 | 808 | |||
AK161558.1 | - | 3 | 5173 | |||
BC145453.1 | - | 7 | 1107 | 9054011 |