Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271704.1 | NP_001258633.1 | 2 | 949 | ||
NM_181317.4 | NP_851834.1 | 2 | 1317 | |||
GenBank mRNA | AK048819.1 | - | 2 | 1291 | ||
AK087481.1 | - | 2 | 997 | |||
AK173110.1 | - | 2 | 1317 | |||
BC059833.1 | - | 2 | 949 | 6838254 |