Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008437.1 | NP_032463.1 | 3 | 486 | ||
GenBank mRNA | AB038144.1 | - | 3 | 499 | ||
AK002520.1 | - | 3 | 333 | |||
AK007861.1 | - | 3 | 332 | |||
AK152663.1 | - | 3 | 324 | |||
AK170013.1 | - | 3 | 330 | |||
BC014813.1 | - | 3 | 319 | 4221844 | ||
D88899.1 | - | 3 | 486 |