Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010616.2 | NP_034746.1 | 6, 7 | 614 | ||
GenBank mRNA | AB001428.1 | - | 2, 3 | 191 | ||
AB755406.1 | - | 6, 7 | 627 | |||
AK018598.1 | - | 6, 7 | 616 | |||
BC022225.1 | - | 6, 7 | 544 | 4222522 |