Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010617.2 | NP_034747.2 | 29 | 3681 | ||
GenBank mRNA | AB037923.2 | - | 29 | 3682 | ||
AK029721.1 | - | 11 | 1317 | |||
AK220490.1 | - | 6 | 635 |