Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008446.2 | NP_032472.2 | 6, 7 | 1192 | ||
GenBank mRNA | AK084837.1 | - | 6, 7 | 1169 | ||
AK135179.1 | - | 6, 7 | 1140 | |||
AK135430.1 | - | 6, 7 | 686 | |||
AK171938.1 | - | 6, 7 | 1105 | |||
BC050946.1 | - | 6, 7 | 1130 | 6822283 | ||
BC054537.1 | - | 6, 7 | 1114 | |||
D12646.1 | - | 6, 7 | 1519 |