Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008446.2 | NP_032472.2 | 16 | 2224 | ||
GenBank mRNA | AK135430.1 | - | 16 | 1718 | ||
AK171938.1 | - | 16 | 2136 | |||
BC050946.1 | - | 16 | 2162 | 6822283 | ||
BC054537.1 | - | 16 | 2146 | |||
D12646.1 | - | 16 | 2551 |