Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008449.2 | NP_032475.2 | 3 | 584 | ||
GenBank mRNA | AB093244.1 | - | 3 | 477 | ||
AF067180.1 | - | 3 | 236 | |||
AK158884.1 | - | 3 | 570 | |||
BC067051.1 | - | 3 | 584 | 30536079 |