Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 4 | 812 | ||
NM_001145816.2 | NP_001139288.1 | 4 | 812 | |||
NM_001256301.1 | NP_001243230.1 | 4 | 542 | |||
NM_001256305.1 | NP_001243234.1 | 4 | 812 | |||
NM_001256307.1 | NP_001243236.1 | 3 | 705 | |||
XM_011531907.1 | XP_011530209.1 | 4 | 807 | |||
GenBank mRNA | AK058013.1 | - | 4 | 401 | ||
AK296642.1 | - | 4 | 542 | |||
AK300524.1 | - | 3 | 448 | |||
AK300940.1 | - | 4 | 401 | |||
AK314624.1 | - | 4 | 401 | |||
BC018986.2 | - | 4 | 374 | 3638799 | ||
J05594.1 | - | 4 | 382 | |||
L76465.1 | - | 4 | 382 | |||
U63296.1 | - | 4 | 365 | |||
X82460.1 | - | 4 | 365 |