Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025280.2 | NP_079556.1 | 3, 4 | 281 | ||
GenBank mRNA | AK009429.1 | - | 3, 4 | 313 | ||
AK132548.1 | - | 3, 4 | 711 | |||
AK167740.1 | - | 3, 4 | 285 | |||
BC028860.1 | - | 3, 4 | 281 | 3673830 | ||
BC058169.1 | - | 3, 4 | 310 | |||
X58472.1 | - | 3, 4 | 286 |