Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013823.2 | NP_038851.2 | 2 | 1323 | ||
GenBank mRNA | AB005141.1 | - | 2 | 1231 | ||
AB010088.1 | - | 2 | 1226 | |||
BC138258.1 | - | 2 | 1323 | 8861277 | ||
BC138259.1 | - | 2 | 1323 | 8861278 |