Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025358.2 | NP_001020529.2 | 12 | 1575 | ||
NM_001025359.2 | NP_001020530.2 | 12 | 1575 | |||
NM_001025360.2 | NP_001020531.2 | 12 | 1594 | |||
NM_001025361.2 | NP_001020532.2 | 12 | 1575 | |||
NM_001025362.2 | NP_001020533.2 | 12 | 1575 | |||
NM_001025363.2 | NP_001020534.2 | 12 | 1575 | |||
NM_001081959.1 | NP_001075428.1 | 12 | 1594 | |||
NM_008450.2 | NP_032476.2 | 12 | 1594 | |||
GenBank mRNA | AF055665.1 | - | 12 | 1490 | ||
AK014256.1 | - | 4 | 253 | |||
AK031309.1 | - | 12 | 1566 | |||
AY753300.1 | - | 11 | 1399 | |||
BC055744.1 | - | 12 | 1430 | 6415280 | ||
CK032061.1 | - | 6 | 499 |