Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 6 | 1107 | ||
GenBank mRNA | AF055666.1 | - | 6 | 940 | ||
AK141439.1 | - | 6 | 1037 | |||
BC014845.1 | - | 6 | 951 | 2599082 | ||
BM963360.1 | - | 2 | 278 |