Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010637.3 | NP_034767.2 | 5 | 1943 | ||
GenBank mRNA | AK141244.1 | - | 3 | 855 | ||
AK144942.1 | - | 5 | 1941 | |||
U70662.1 | - | 5 | 1937 |