Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020995.3 | NP_066275.3 | 3, 4 | 234 | ||
GenBank mRNA | AV698541.1 | - | 3, 4 | 219 | ||
BM547990.1 | - | 1, 2 | 121 | |||
CB115435.1 | - | 3, 4 | 233 | |||
CB147217.1 | - | 3, 4 | 234 | |||
T71114.1 | - | 3, 4 | 219 |
Gene Symbol: HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318138.1 | NP_001305067.1 | 3, 4 | 262 | ||
NM_005143.4 | NP_005134.1 | 3, 4 | 262 | |||
GenBank mRNA | AK314700.1 | - | 3, 4 | 231 | ||
BC121124.1 | - | 3, 4 | 225 | 40120682 | ||
BC121125.1 | - | 3, 4 | 225 | 40120683 | ||
CB117006.1 | - | 3, 4 | 244 | |||
CB134891.1 | - | 3, 4 | 245 | |||
L29394.1 | - | 3, 4 | 227 |