Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010643.1 | NP_034773.1 | 291 | |||
GenBank mRNA | AB039277.1 | - | 291 | |||
BC146015.1 | - | 3 | 318 | 40131318 | ||
BC146019.1 | - | 3 | 318 | 40131322 |
Gene Symbol: Klk1b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020268.3 | NP_064664.1 | 3 | 332 | ||
GenBank mRNA | AB039275.1 | - | 3 | 334 | ||
BC132090.1 | - | 3 | 309 | 40130367 | ||
BC132665.1 | - | 3 | 309 | 40130942 |