Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010642.2 | NP_034772.1 | 5 | 769 | ||
GenBank mRNA | AB039276.1 | - | 5 | 772 | ||
AK144221.1 | - | 5 | 736 | |||
BC012243.1 | - | 5 | 703 | 4167772 | ||
BC145383.1 | - | 5 | 725 | 9053941 |
Gene Symbol: Klk1b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020268.3 | NP_064664.1 | 5 | 754 | ||
GenBank mRNA | AB039275.1 | - | 5 | 756 | ||
BC132090.1 | - | 5 | 731 | 40130367 | ||
BC132665.1 | - | 5 | 731 | 40130942 |