Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008456.3 | NP_032482.1 | 5 | 769 | ||
GenBank mRNA | BC013659.1 | - | 5 | 763 | 4190286 | |
BC094281.1 | - | 5 | 769 | 4923171 |