Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010645.2 | NP_034775.1 | 3 | 291 | ||
GenBank mRNA | AK089370.1 | - | 3 | 309 | ||
BC026378.1 | - | 3 | 291 | 4165617 |