Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008458.2 | NP_032484.1 | 5 | 1318 | ||
GenBank mRNA | BC116724.1 | - | 5 | 1316 | 40106394 |