Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252577.1 | NP_001239506.1 | 4 | 498 | ||
NM_010649.3 | NP_034779.1 | 4 | 501 | |||
GenBank mRNA | AK080171.1 | - | 4 | 499 | ||
AK145141.1 | - | 3 | 369 | |||
AY860975.1 | - | 3 | 272 | |||
L78247.1 | - | 3 | 353 | |||
U10090.1 | - | 3 | 411 |