Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008463.2 | NP_032489.1 | 2 | 168 | ||
GenBank mRNA | AF247643.1 | - | 2 | 137 | ||
AF419251.1 | - | 301 | ||||
BC120550.1 | - | 2 | 126 | 40129473 | ||
BC131790.1 | - | 2 | 110 | 40101177 | ||
BC137708.1 | - | 2 | 126 | 8860727 | ||
FJ159531.1 | - | 1 | 60 | |||
U10091.1 | - | 2 | 168 |