Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 9, 10 | 1074 | ||
GenBank mRNA | AK028259.1 | - | 9, 10 | 1075 | ||
AK028307.1 | - | 9, 10 | 1093 | |||
AK154800.1 | - | 9, 10 | 1092 | |||
AK169311.1 | - | 9, 10 | 1097 | |||
AK169348.1 | - | 9, 10 | 1097 | |||
BC006771.1 | - | 9, 10 | 1059 | 3588534 | ||
U20619.1 | - | 9, 10 | 1079 | |||
U34228.1 | - | 8, 9 | 923 |