Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010655.3 | NP_034785.1 | 10 | 1621 | ||
GenBank mRNA | AK075952.1 | - | 10 | 1561 | ||
AK088496.1 | - | 10 | 1560 | |||
AK139850.1 | - | 10 | 1558 | |||
AK147135.1 | - | 10 | 1560 | |||
AK165577.1 | - | 10 | 1562 | |||
AK166577.1 | - | 10 | 1564 | |||
AK166790.1 | - | 10 | 1560 | |||
AK166982.1 | - | 10 | 1558 | |||
AK167551.1 | - | 10 | 1560 | |||
AK169283.1 | - | 10 | 1560 | |||
BC003274.1 | - | 10 | 1543 | 3499880 | ||
BC006720.1 | - | 10 | 1541 | 3711035 | ||
BC082280.1 | - | 9 | 1426 | 6334979 | ||
BC086789.1 | - | 10 | 1536 | |||
BC094011.1 | - | 10 | 1533 | 5035738 | ||
BC106110.1 | - | 10 | 1533 | |||
D55720.1 | - | 10 | 1608 | |||
U12270.1 | - | 10 | 1546 | |||
U34229.1 | - | 9 | 1431 |