Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008466.5 | NP_032492.1 | 7 | 575 | ||
GenBank mRNA | AF020772.1 | - | 7 | 419 | ||
AK011559.1 | - | 7 | 446 | |||
AK049373.1 | - | 7 | 446 | |||
AK161664.1 | - | 7 | 504 | |||
BC026885.1 | - | 7 | 469 | 3979788 |