Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010656.2 | NP_034786.1 | 3 | 532 | ||
GenBank mRNA | AK148179.1 | - | 3 | 534 | ||
AK162864.1 | - | 3 | 557 | |||
AK163041.1 | - | 3 | 558 | |||
AK164430.1 | - | 3 | 558 | |||
AK164529.1 | - | 3 | 504 | |||
AK188742.1 | - | 1 | 75 | |||
BC021484.1 | - | 3 | 557 | 5344514 | ||
U02487.1 | - | 3 | 532 |