Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010662.1 | NP_034792.1 | 2, 3 | 556 | ||
GenBank mRNA | AK004635.1 | - | 2, 3 | 628 | ||
AK009677.1 | - | 2, 3 | 117 | |||
AK010156.1 | - | 2, 3 | 628 | |||
AK132640.1 | - | 2, 3 | 628 | |||
BC060286.1 | - | 2, 3 | 571 | 30292772 | ||
X03492.1 | - | 2, 3 | 181 |