Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008469.2 | NP_032495.2 | 8 | 1407 | ||
GenBank mRNA | BC057934.1 | - | 8 | 1386 | 5251453 | |
CA451823.1 | - | 4 | 414 |