Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008470.1 | NP_032496.1 | 7 | 1316 | ||
GenBank mRNA | AF053235.1 | - | 7 | 1316 | ||
AK137631.1 | - | 7 | 1380 | |||
BC103615.1 | - | 7 | 1319 | 40046209 | ||
BC103616.1 | - | 7 | 1319 | 40046211 | ||
BC103617.1 | - | 7 | 1319 | 40046210 | ||
BC103666.1 | - | 7 | 1316 | 40046212 | ||
BC114977.1 | - | 7 | 1316 | 40046220 |