Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008474.2 | NP_032500.2 | 2 | 730 | ||
GenBank mRNA | AY028607.1 | - | 2 | 687 | ||
BC114971.1 | - | 2 | 683 | 40039508 | ||
CF173770.1 | - | 2 | 464 |