Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008475.2 | NP_032501.2 | 6 | 1150 | ||
GenBank mRNA | BC064008.1 | - | 6 | 1131 | ||
BC145839.1 | - | 6 | 1105 | 40131142 | ||
BC145841.1 | - | 6 | 1105 | 40131144 | ||
X03491.1 | - | 6 | 1202 |