Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008476.3 | NP_032502.3 | 3 | 825 | ||
GenBank mRNA | AK009394.1 | - | 3 | 828 | ||
BC064470.1 | - | 3 | 799 | |||
BC080820.1 | - | 3 | 825 | 30141946 | ||
K02108.1 | - | 3 | 817 |