Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap6-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010673.2 | NP_034803.2 | 1 | 544 | ||
GenBank mRNA | AA763314.1 | - | 1 | 39 | ||
BC116356.1 | - | 1 | 543 | |||
BC116357.1 | - | 1 | 543 | |||
BC128485.1 | - | 1 | 543 | |||
BC128486.1 | - | 1 | 543 | |||
D89902.1 | - | 2 | 556 |