Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ktn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008477.2 | NP_032503.2 | 38, 39 | 3727 | ||
GenBank mRNA | AJ517368.1 | - | 14, 15 | 1065 | ||
AJ517369.1 | - | 14, 15 | 1063 | |||
AJ517371.1 | - | 16, 17 | 1203 | |||
AJ517373.1 | - | 15, 16 | 1150 | |||
AJ517376.1 | - | 15, 16 | 1135 | |||
AJ517377.1 | - | 16, 17 | 1222 | |||
DT913164.1 | - | 4, 5 | 298 | |||
L43326.1 | - | 38, 39 | 3715 |