Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lama3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010680.1 | NP_034810.1 | 35 | 4564 | ||
GenBank mRNA | X84014.1 | - | 16 | 2282 |