Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008485.3 | NP_032511.3 | 19 | 2988 | ||
GenBank mRNA | DV654715.1 | - | 2 | 101 | ||
U43327.2 | - | 18 | 2718 |