Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011029.4 | NP_035159.3 | 7 | 1378 | ||
GenBank mRNA | AK082935.1 | - | 7 | 1358 |