Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008486.2 | NP_032512.2 | 1 | 326 | ||
GenBank mRNA | AK143746.1 | - | 1 | 269 | ||
AK171427.1 | - | 2 | 686 | |||
BC005431.1 | - | 1 | 292 | 3593307 | ||
U77083.1 | - | 1 | 258 |