Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010686.3 | NP_034816.1 | 2 | 159 | ||
GenBank mRNA | AF364050.1 | - | 2 | 177 | ||
AK031802.1 | - | 2 | 173 | |||
AK050633.1 | - | 2 | 174 | |||
AK089432.1 | - | 2 | 209 | |||
AK089508.1 | - | 2 | 182 | |||
AK089535.1 | - | 2 | 174 | |||
AK150299.1 | - | 2 | 170 | |||
AK152918.1 | - | 2 | 170 | |||
AK153877.1 | - | 2 | 173 | |||
AK155744.1 | - | 2 | 167 | |||
AK163146.1 | - | 2 | 174 | |||
AK170438.1 | - | 2 | 161 | |||
AK170690.1 | - | 2 | 193 | |||
AK170867.1 | - | 2 | 177 | |||
AK171053.1 | - | 2 | 184 | |||
AK171057.1 | - | 2 | 209 | |||
AK171105.1 | - | 2 | 174 | |||
AK171116.1 | - | 2 | 174 | |||
AK171149.1 | - | 2 | 182 | |||
AK171172.1 | - | 174 | ||||
AK171193.1 | - | 2 | 174 | |||
AK171213.1 | - | 2 | 174 | |||
AK171236.1 | - | 2 | 174 | |||
AK171244.1 | - | 2 | 174 | |||
AK171250.1 | - | 2 | 195 | |||
AK171269.1 | - | 2 | 211 | |||
AK171270.1 | - | 2 | 177 | |||
AK171274.1 | - | 2 | 167 | |||
AK171280.1 | - | 2 | 207 | |||
AK171285.1 | - | 2 | 211 | |||
AK171481.1 | - | 2 | 210 | |||
AK171547.1 | - | 2 | 174 | |||
AK171562.1 | - | 2 | 165 | |||
AK171571.1 | - | 2 | 191 | |||
AK171575.1 | - | 2 | 209 | |||
AK172306.1 | - | 2 | 174 | |||
AK172486.1 | - | 2 | 209 | |||
AK172707.1 | - | 2 | 174 | |||
AK172885.1 | - | 2 | 149 | |||
BC020993.1 | - | 2 | 159 | 4037686 | ||
BC055057.1 | - | 2 | 146 | 6529760 | ||
BC114403.1 | - | 2 | 139 | |||
BC158032.1 | - | 2 | 355 | 9007335 | ||
U29539.1 | - | 2 | 177 | |||
U51239.1 | - | 2 | 198 |
Gene Symbol: Ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AX775991.1 | - | 169 |