Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 25, 26 | 2595 | ||
NM_001130151.1 | NP_001123623.1 | 24, 25 | 2586 | |||
NM_001130152.1 | NP_001123624.1 | 23, 24 | 2427 | |||
NM_001130153.1 | NP_001123625.1 | 23, 24 | 2418 | |||
NM_008488.2 | NP_032514.1 | 23, 24 | 2415 | |||
GenBank mRNA | AK012156.1 | - | 23, 24 | 2369 | ||
AK043027.1 | - | 24, 25 | 2551 | |||
AK051518.1 | - | 23, 24 | 2369 | |||
AK131181.1 | - | 23, 24 | 2315 | |||
AK157056.1 | - | 25, 26 | 2531 | |||
AK172354.1 | - | 25, 26 | 2531 | |||
AK218306.1 | - | 1, 2 | 83 | |||
AY246272.1 | - | 2, 3 | 250 | |||
AY246273.1 | - | 2, 3 | 250 | |||
BC012488.1 | - | 23, 24 | 2378 | 3154558 | ||
U58203.1 | - | 23, 24 | 2345 |