Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 30 | 3010 | ||
NM_001130151.1 | NP_001123623.1 | 29 | 3001 | |||
NM_001130152.1 | NP_001123624.1 | 28 | 2842 | |||
NM_001130153.1 | NP_001123625.1 | 28 | 2833 | |||
NM_008488.2 | NP_032514.1 | 28 | 2830 | |||
GenBank mRNA | AK012156.1 | - | 28 | 2784 | ||
AK043027.1 | - | 29 | 2966 | |||
AK051518.1 | - | 28 | 2785 | |||
AK131181.1 | - | 28 | 2730 | |||
AK157056.1 | - | 30 | 2946 | |||
AK172354.1 | - | 30 | 2946 | |||
AK203844.1 | - | 2 | 136 | |||
AY246272.1 | - | 6 | 501 | |||
BC012488.1 | - | 28 | 2793 | 3154558 | ||
U58203.1 | - | 28 | 2760 |