Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010695.1 | NP_034825.1 | 4 | 410 | ||
GenBank mRNA | BC114544.1 | - | 4 | 409 | 40050061 | |
D38581.1 | - | 4 | 410 |