Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013581.3 | NP_038609.3 | 7, 8 | 2091 | ||
GenBank mRNA | AF109377.2 | - | 7, 8 | 2074 | ||
AK034636.2 | - | 7, 8 | 2096 | |||
AK152464.1 | - | 7, 8 | 2082 | |||
AK158507.1 | - | 7, 8 | 2453 | |||
AK159497.1 | - | 7, 8 | 2093 | |||
AK162971.1 | - | 7, 8 | 2092 | |||
AK171574.1 | - | 7, 8 | 2085 | |||
AK185855.1 | - | 1, 2 | 19 | |||
BC063056.1 | - | 7, 8 | 2073 | 5720460 |