Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252658.1 | NP_001239587.1 | 12 | 1935 | ||
NM_001252659.1 | NP_001239588.1 | 12 | 1932 | |||
NM_010700.3 | NP_034830.2 | 12 | 1935 | |||
GenBank mRNA | AF425607.1 | - | 12 | 1783 | ||
AK149950.1 | - | 12 | 1873 | |||
AK151968.1 | - | 12 | 1902 | |||
AK152103.1 | - | 12 | 1871 | |||
AK160006.1 | - | 12 | 1873 | |||
AK166466.1 | - | 12 | 1872 | |||
AK170264.1 | - | 12 | 1880 | |||
BC019207.1 | - | 12 | 1853 | 5052599 | ||
BC053041.1 | - | 12 | 1880 | 5704871 | ||
X64414.1 | - | 12 | 1854 | |||
Z19521.1 | - | 12 | 1733 |