Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252658.1 | NP_001239587.1 | 11 | 1867 | ||
NM_001252659.1 | NP_001239588.1 | 11 | 1864 | |||
NM_010700.3 | NP_034830.2 | 11 | 1867 | |||
GenBank mRNA | AF425607.1 | - | 11 | 1715 | ||
AK149950.1 | - | 11 | 1805 | |||
AK151968.1 | - | 11 | 1834 | |||
AK152103.1 | - | 11 | 1803 | |||
AK160006.1 | - | 11 | 1805 | |||
AK166466.1 | - | 11 | 1804 | |||
AK170264.1 | - | 11 | 1812 | |||
BC019207.1 | - | 11 | 1785 | 5052599 | ||
BC053041.1 | - | 11 | 1812 | 5704871 | ||
X64414.1 | - | 11 | 1786 | |||
Z19521.1 | - | 11 | 1665 |